Product Details

SNP ID
rs61431871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8368472 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTTACACTGTCACAAGCCAAGG[A/C]CTGCCAAGGATTGCCGATGGCCACT
Phenotype
MIM: 603704
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KRBA2 PubMed Links

Gene Details

Gene
KRBA2
Gene Name
KRAB-A domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304947.1 3207 Intron NP_001291876.1
NM_213597.3 3207 Intron NP_998762.1
XM_005256443.2 3207 Intron XP_005256500.1
XM_011523643.1 3207 Intron XP_011521945.1
XM_011523644.1 3207 Intron XP_011521946.1
XM_017024163.1 3207 Intron XP_016879652.1
XM_017024164.1 3207 UTR 3 XP_016879653.1
XM_017024165.1 3207 UTR 3 XP_016879654.1
Gene
LOC100128288
Gene Name
uncharacterized LOC100128288
There are no transcripts associated with this gene.

Gene
RPL26
Gene Name
ribosomal protein L26
There are no transcripts associated with this gene.

View Full Product Details