Product Details

SNP ID
rs62077531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39606022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGTGGGCTGCGACAGACCCTTG[C/T]ACAGAGTCTGCACGTAGGACACTAG
Phenotype
MIM: 601725
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NEUROD2 PubMed Links

Gene Details

Gene
NEUROD2
Gene Name
neuronal differentiation 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006160.3 778 Missense Mutation TAC,TGC Y193C NP_006151.3

View Full Product Details