Product Details

SNP ID
rs56226305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41315521 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTGCCCCCGCAGCCAGAGCCCC[C/T]GCAGCCAGAGCCCCCACAGCCACAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP16-1 PubMed Links

Gene Details

Gene
KRTAP16-1
Gene Name
keratin associated protein 16-1
There are no transcripts associated with this gene.

Gene
KRTAP17-1
Gene Name
keratin associated protein 17-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031964.1 175 Missense Mutation AGG,GGG R44G NP_114170.1

View Full Product Details