Product Details

SNP ID
hCV89349955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43000094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGTATAACTACAATCACCTAAT[A/G]CCCACAAGGTGAGCATTTCAAGAAC
Phenotype
MIM: 600735 MIM: 607526
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFI35 PubMed Links

Gene Details

Gene
IFI35
Gene Name
interferon induced protein 35
There are no transcripts associated with this gene.

Gene
RPL27
Gene Name
ribosomal protein L27
There are no transcripts associated with this gene.

Gene
RUNDC1
Gene Name
RUN domain containing 1
There are no transcripts associated with this gene.

View Full Product Details