Product Details

SNP ID
rs59789372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38731717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCTGTTTGTTTCTCAGAGGTGC[A/G]CTGCATGGCGAAGCTCACCTGGCCC
Phenotype
MIM: 611933 MIM: 600328 MIM: 600346
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CISD3 PubMed Links

Gene Details

Gene
CISD3
Gene Name
CDGSH iron sulfur domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136498.1 Intron NP_001129970.1
Gene
MLLT6
Gene Name
MLLT6, PHD finger domain containing
There are no transcripts associated with this gene.

Gene
PCGF2
Gene Name
polycomb group ring finger 2
There are no transcripts associated with this gene.

View Full Product Details