Product Details

SNP ID
hCV89354750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58694935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCTTATTTTACTTTCAGAAGT[G/T]GGGATATCTAAAGCAGAAGCCTTAG
Phenotype
MIM: 602774 MIM: 605792
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RAD51C PubMed Links

Gene Details

Gene
RAD51C
Gene Name
RAD51 paralog C
There are no transcripts associated with this gene.

Gene
TEX14
Gene Name
testis expressed 14, intercellular bridge forming factor
There are no transcripts associated with this gene.

View Full Product Details