Product Details

SNP ID
rs58325641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:60444947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCACCTCACCATATGTAAAAAGTT[C/T]GCGCAAGTTAACTTATTGTGAGATA
Phenotype
MIM: 605324
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APPBP2 PubMed Links

Gene Details

Gene
APPBP2
Gene Name
amyloid beta precursor protein binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282476.1 3913 UTR 3 NP_001269405.1
NM_006380.3 3913 UTR 3 NP_006371.2
XM_011524198.2 3913 UTR 3 XP_011522500.1
XM_011524199.2 3913 UTR 3 XP_011522501.1

View Full Product Details