Product Details

SNP ID
hCV89367557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50097432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTTCTCCTGCCAGGATCCAGCA[A/G]TGCCTGTGAGAAAACCTCCTTCACC
Phenotype
MIM: 605025 MIM: 602525
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGA3 PubMed Links

Gene Details

Gene
ITGA3
Gene Name
integrin subunit alpha 3
There are no transcripts associated with this gene.

Gene
PDK2
Gene Name
pyruvate dehydrogenase kinase 2
There are no transcripts associated with this gene.

View Full Product Details