Product Details
- SNP ID
-
rs59168537
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:44036279 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTTGCAGTATCTTTTGGCTTTCCA[C/G]GTCTCTAAAATGTTTTTCAACCTGA
- Phenotype
-
MIM: 611793
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LSM12
PubMed Links
Gene Details
- Gene
- LSM12
- Gene Name
- LSM12 homolog
View Full Product Details