Product Details

SNP ID
rs55800471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:13665767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTCACAACAGAGGTTTATGTTTG[A/G]GGTGATCACCACTAATACACACTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM210A PubMed Links

Gene Details

Gene
FAM210A
Gene Name
family with sequence similarity 210 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098801.1 1824 UTR 3 NP_001092271.1
NM_152352.3 1824 UTR 3 NP_689565.2

View Full Product Details