Product Details

SNP ID
hCV89392946
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:9887848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGTGAAAGTGATCCTGAGCAAG[A/G]AGGACTTTGAGGCATCACTGAAGGA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TXNDC2 PubMed Links

Gene Details

Gene
TXNDC2
Gene Name
thioredoxin domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098529.1 1818 Missense Mutation AAG,GAG K457E NP_001091999.1
NM_032243.5 1818 Missense Mutation AAG,GAG K390E NP_115619.4

View Full Product Details