Product Details

SNP ID
rs57772091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:23139621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCAATGATTACACCACCTCTGT[C/T]CACTGGACTGGATTCTTGACATCTA
Phenotype
MIM: 609194
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CABLES1 PubMed Links

Gene Details

Gene
CABLES1
Gene Name
Cdk5 and Abl enzyme substrate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100619.2 Intron NP_001094089.1
NM_001256438.1 Intron NP_001243367.1
NM_138375.2 Intron NP_612384.1
Gene
LOC105372022
Gene Name
uncharacterized LOC105372022
There are no transcripts associated with this gene.

View Full Product Details