Product Details
- SNP ID
-
rs61760984
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:46812517 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACTCCTGCTGCTGGATGAACTAAC[A/G]TGAGTACTGCTTTCATGGGAGCTGG
- Phenotype
-
MIM: 603567
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PIAS2
PubMed Links
Gene Details
- Gene
- PIAS2
- Gene Name
- protein inhibitor of activated STAT 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001324046.1 |
2271 |
UTR 3 |
|
|
NP_001310975.1 |
NM_001324047.1 |
2271 |
Silent Mutation |
CAC,CAT |
H586H |
NP_001310976.1 |
NM_001324048.1 |
2271 |
Silent Mutation |
CAC,CAT |
H585H |
NP_001310977.1 |
NM_001324049.1 |
2271 |
Silent Mutation |
CAC,CAT |
H585H |
NP_001310978.1 |
NM_001324051.1 |
2271 |
Intron |
|
|
NP_001310980.1 |
NM_001324052.1 |
2271 |
Intron |
|
|
NP_001310981.1 |
NM_001324053.1 |
2271 |
Intron |
|
|
NP_001310982.1 |
NM_001324054.1 |
2271 |
Intron |
|
|
NP_001310983.1 |
NM_001324055.1 |
2271 |
Intron |
|
|
NP_001310984.1 |
NM_001324057.1 |
2271 |
Intron |
|
|
NP_001310986.1 |
NM_001324058.1 |
2271 |
Intron |
|
|
NP_001310987.1 |
NM_001324059.1 |
2271 |
Intron |
|
|
NP_001310988.1 |
NM_001324060.1 |
2271 |
Intron |
|
|
NP_001310989.1 |
NM_004671.4 |
2271 |
Silent Mutation |
CAC,CAT |
H594H |
NP_004662.2 |
NM_173206.3 |
2271 |
Intron |
|
|
NP_775298.1 |
XM_005258377.3 |
2271 |
Silent Mutation |
CAC,CAT |
H599H |
XP_005258434.1 |
XM_005258379.3 |
2271 |
Silent Mutation |
CAC,CAT |
H595H |
XP_005258436.1 |
XM_006722571.3 |
2271 |
Silent Mutation |
CAC,CAT |
H634H |
XP_006722634.1 |
XM_006722572.3 |
2271 |
Silent Mutation |
CAC,CAT |
H633H |
XP_006722635.1 |
XM_006722573.2 |
2271 |
Intron |
|
|
XP_006722636.1 |
XM_011526258.2 |
2271 |
Silent Mutation |
CAC,CAT |
H593H |
XP_011524560.1 |
XM_011526259.2 |
2271 |
Silent Mutation |
CAC,CAT |
H586H |
XP_011524561.1 |
XM_017026068.1 |
2271 |
Silent Mutation |
CAC,CAT |
H586H |
XP_016881557.1 |
XM_017026069.1 |
2271 |
Silent Mutation |
CAC,CAT |
H585H |
XP_016881558.1 |
XM_017026070.1 |
2271 |
Silent Mutation |
CAC,CAT |
H585H |
XP_016881559.1 |
XM_017026071.1 |
2271 |
Intron |
|
|
XP_016881560.1 |
XM_017026072.1 |
2271 |
Intron |
|
|
XP_016881561.1 |
View Full Product Details