Product Details

SNP ID
rs56093302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:51175721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATCCACTGATCATACCAGCCTT[A/G]AGCACTCTGGGCGAGATGAAAGTTG
Phenotype
MIM: 611005
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MEX3C PubMed Links

Gene Details

Gene
MEX3C
Gene Name
mex-3 RNA binding family member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016626.4 2971 UTR 3 NP_057710.3

View Full Product Details