Product Details

SNP ID
hCV89421526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38405616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGACCTCACTACATCTGCTTTCC[A/C]CTGGAGTGACCGGGCAGAGGTGTGG
Phenotype
MIM: 607320 MIM: 609293
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM98C PubMed Links

Gene Details

Gene
FAM98C
Gene Name
family with sequence similarity 98 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174905.3 750 Missense Mutation CAC,CCC H244P NP_777565.3
Gene
RASGRP4
Gene Name
RAS guanyl releasing protein 4
There are no transcripts associated with this gene.

Gene
SPRED3
Gene Name
sprouty related EVH1 domain containing 3
There are no transcripts associated with this gene.

View Full Product Details