Product Details

SNP ID
rs62118220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44618371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAGCTACACCGTCGTGCTGGAA[A/G]CCAGCAGGGGACACAGGAGTGTGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGSF23 PubMed Links

Gene Details

Gene
IGSF23
Gene Name
immunoglobulin superfamily member 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205280.1 862 Intron NP_001192209.1
XM_017026317.1 862 Intron XP_016881806.1
XM_017026318.1 862 Intron XP_016881807.1
XM_017026319.1 862 Intron XP_016881808.1
XM_017026320.1 862 Missense Mutation ACC,GCC T15A XP_016881809.1
XM_017026321.1 862 Missense Mutation ACC,GCC T15A XP_016881810.1
XM_017026322.1 862 Missense Mutation ACC,GCC T15A XP_016881811.1
XM_017026323.1 862 Intron XP_016881812.1
XM_017026324.1 862 Intron XP_016881813.1
XM_017026325.1 862 Intron XP_016881814.1
XM_017026326.1 862 Missense Mutation ACC,GCC T15A XP_016881815.1

View Full Product Details