Product Details

SNP ID
rs57398651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51936110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACCTGGATACCATCCTTTGCAG[C/G]GATGTAATTCACCAGAGACCAAGAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF613 PubMed Links

Gene Details

Gene
ZNF613
Gene Name
zinc finger protein 613
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031721.3 570 UTR 5 NP_001026891.2
NM_024840.3 570 Intron NP_079116.2
XM_005259269.3 570 UTR 5 XP_005259326.1
XM_011527333.2 570 UTR 5 XP_011525635.1
XM_017027315.1 570 Intron XP_016882804.1

View Full Product Details