Product Details

SNP ID
rs59773005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56595959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATGAATGTGAATGTGTCCATGT[A/G]TGTCCGTGTGGGTCATGTGTGGTAG
Phenotype
MIM: 194545
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF71 PubMed Links

Gene Details

Gene
ZNF71
Gene Name
zinc finger protein 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021216.4 341 Intron NP_067039.1
XM_011527193.2 341 Intron XP_011525495.1
XM_017027078.1 341 Intron XP_016882567.1
XM_017027079.1 341 UTR 5 XP_016882568.1
XM_017027080.1 341 Intron XP_016882569.1

View Full Product Details