Product Details

SNP ID
rs55976703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35294475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGACTGCAGACCAACTGTGTGGC[C/T]TTGGGCGAGTCACTTAACCTCTCTG
Phenotype
MIM: 606464 MIM: 159460
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HAMP PubMed Links

Gene Details

Gene
HAMP
Gene Name
hepcidin antimicrobial peptide
There are no transcripts associated with this gene.

Gene
MAG
Gene Name
myelin associated glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199216.1 Intron NP_001186145.1
NM_002361.3 Intron NP_002352.1
NM_080600.2 Intron NP_542167.1

View Full Product Details