Product Details

SNP ID
rs57373049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:591355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGGGCGCGAGCCTGTGGCCGGA[A/G]AGGTGTGTCTCCAGGAGTGTGTTTG
Phenotype
MIM: 109480 MIM: 602781
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BSG PubMed Links

Gene Details

Gene
BSG
Gene Name
basigin (Ok blood group)
There are no transcripts associated with this gene.

Gene
HCN2
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001194.3 Intron NP_001185.3

View Full Product Details