Product Details

SNP ID
hCV89460555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5693313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACACTCACCGCAATGGTCTTCTC[A/C]TTGATGCCACCAACAGGCAGGATCT
Phenotype
MIM: 605490
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HSD11B1L PubMed Links

Gene Details

Gene
HSD11B1L
Gene Name
hydroxysteroid 11-beta dehydrogenase 1 like
There are no transcripts associated with this gene.

Gene
LONP1
Gene Name
lon peptidase 1, mitochondrial
There are no transcripts associated with this gene.

Gene
RPL36
Gene Name
ribosomal protein L36
There are no transcripts associated with this gene.

View Full Product Details