Product Details

SNP ID
rs61418834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19529094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATAGCTGGGACAGAATGGGGTC[C/T]GGGGACTGGAGACGGGCACAGCCGG
Phenotype
MIM: 612419 MIM: 609435
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CILP2 PubMed Links

Gene Details

Gene
CILP2
Gene Name
cartilage intermediate layer protein 2
There are no transcripts associated with this gene.

Gene
NDUFA13
Gene Name
NADH:ubiquinone oxidoreductase subunit A13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015965.6 194 Intron NP_057049.5
Gene
YJEFN3
Gene Name
YjeF N-terminal domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190328.1 194 Intron NP_001177257.1
NM_198537.3 194 Intron NP_940939.2
XM_005259903.4 194 Silent Mutation TCC,TCT S3S XP_005259960.1
XM_011527996.2 194 Silent Mutation TCC,TCT S3S XP_011526298.1
XM_011527997.1 194 Intron XP_011526299.1
XM_011527998.2 194 Intron XP_011526300.1
XM_011527999.1 194 Intron XP_011526301.1
XM_017026770.1 194 Intron XP_016882259.1
XM_017026771.1 194 Intron XP_016882260.1

View Full Product Details