Product Details

SNP ID
rs56872664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9757695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTGCATATTAAGATTTGTGGAA[C/G]GAGCAAATGCTTTACCGCATTCCTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF846 PubMed Links

Gene Details

Gene
ZNF846
Gene Name
zinc finger protein 846
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077624.1 2353 Missense Mutation CCT,CGT P461R NP_001071092.1
XM_005259772.3 2353 Missense Mutation XP_005259829.1
XM_005259773.4 2353 Missense Mutation XP_005259830.1
XM_005259775.3 2353 Missense Mutation XP_005259832.1
XM_006722658.3 2353 Intron XP_006722721.1
XM_011527717.2 2353 Missense Mutation XP_011526019.1
XM_011527718.2 2353 Missense Mutation XP_011526020.1
XM_011527719.2 2353 Missense Mutation XP_011526021.1
XM_011527720.2 2353 Missense Mutation XP_011526022.1
XM_011527721.2 2353 Missense Mutation XP_011526023.1
XM_017026404.1 2353 Missense Mutation XP_016881893.1
XM_017026405.1 2353 Missense Mutation XP_016881894.1
XM_017026406.1 2353 Missense Mutation XP_016881895.1
XM_017026407.1 2353 Intron XP_016881896.1
XM_017026408.1 2353 Intron XP_016881897.1

View Full Product Details