Product Details

SNP ID
hCV89465892
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21423044 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTTAAGAAAAGGATGTAAAAGT[A/G]TGAATGAGTGTAATGTGCACAAAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF493 PubMed Links

Gene Details

Gene
ZNF493
Gene Name
zinc finger protein 493
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076678.2 281 Intron NP_001070146.1
NM_145326.2 281 Intron NP_663299.2
NM_175910.6 281 Missense Mutation ATG,GTG M1V NP_787106.4

View Full Product Details