Product Details

SNP ID
rs62103612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9140582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCACCTCCGCTCCCCCGATCCTA[C/T]TCCCAGTCGTGTAAGCCCTGCTTTC
Phenotype
MIM: 613864
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF317 PubMed Links

Gene Details

Gene
ZNF317
Gene Name
zinc finger protein 317
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190791.1 203 UTR 5 NP_001177720.1
NM_020933.4 203 UTR 5 NP_065984.3

View Full Product Details