Product Details

SNP ID
rs62121155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18104351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCAGACATATTCAAGCAGTCAG[A/G]GAGACCTTCGAGGGGTAGATTATTC
Phenotype
MIM: 601604 MIM: 612258
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL12RB1 PubMed Links

Gene Details

Gene
IL12RB1
Gene Name
interleukin 12 receptor subunit beta 1
There are no transcripts associated with this gene.

Gene
MAST3
Gene Name
microtubule associated serine/threonine kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015016.1 Intron NP_055831.1
XM_005259823.3 Intron XP_005259880.1
XM_005259824.2 Intron XP_005259881.1
XM_005259825.2 Intron XP_005259882.1
XM_005259826.3 Intron XP_005259883.1
XM_005259827.2 Intron XP_005259884.1
XM_006722698.2 Intron XP_006722761.1
XM_006722699.2 Intron XP_006722762.2
XM_006722700.3 Intron XP_006722763.1
XM_011527823.1 Intron XP_011526125.1
XM_017026506.1 Intron XP_016881995.1
XM_017026507.1 Intron XP_016881996.1
XM_017026508.1 Intron XP_016881997.1
XM_017026509.1 Intron XP_016881998.1
XM_017026510.1 Intron XP_016881999.1
XM_017026511.1 Intron XP_016882000.1
XM_017026512.1 Intron XP_016882001.1
XM_017026513.1 Intron XP_016882002.1
XM_017026514.1 Intron XP_016882003.1
XM_017026515.1 Intron XP_016882004.1
XM_017026516.1 Intron XP_016882005.1

View Full Product Details