Product Details

SNP ID
rs59589815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18175171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGGGAGCTCTTCCCAACATGGCT[A/G]TTGGTCATGGAGATCCTCAATGTCA
Phenotype
MIM: 604664 MIM: 603157
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFI30 PubMed Links

Gene Details

Gene
IFI30
Gene Name
IFI30, lysosomal thiol reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006332.4 326 Silent Mutation CTA,CTG L88L NP_006323.2
Gene
PIK3R2
Gene Name
phosphoinositide-3-kinase regulatory subunit 2
There are no transcripts associated with this gene.

View Full Product Details