Product Details

SNP ID
rs57325871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:52084247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGATGGATCAGTTTTAAAATTG[G/T]AATCCGTTACGTAGAAGCTATCCCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZFP64 PubMed Links

Gene Details

Gene
ZFP64
Gene Name
ZFP64 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319146.1 2239 UTR 3 NP_001306075.1
NM_018197.2 2239 Intron NP_060667.2
NM_022088.4 2239 Intron NP_071371.3
NM_199426.1 2239 Intron NP_955458.1
NM_199427.2 2239 UTR 3 NP_955459.2
XM_005260449.1 2239 Intron XP_005260506.1
XM_006723822.1 2239 Intron XP_006723885.1
XM_011528900.1 2239 Intron XP_011527202.1
XM_017027945.1 2239 UTR 3 XP_016883434.1
XM_017027946.1 2239 Intron XP_016883435.1
XM_017027947.1 2239 UTR 3 XP_016883436.1
XM_017027948.1 2239 Intron XP_016883437.1
XM_017027949.1 2239 Intron XP_016883438.1

View Full Product Details