Product Details

SNP ID
rs61558260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58839812 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACCTCTCTCGAGTCTTAGGCTG[C/G]GGAATCTAAGACTCAGCGAGAGGAG
Phenotype
MIM: 139320 MIM: 610540
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GNAS PubMed Links

Gene Details

Gene
GNAS
Gene Name
GNAS complex locus
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000516.5 132 Intron NP_000507.1
NM_001077488.3 132 Intron NP_001070956.1
NM_001077489.3 132 Intron NP_001070957.1
NM_001077490.2 132 Intron NP_001070958.1
NM_001309840.1 132 Intron NP_001296769.1
NM_001309842.1 132 Intron NP_001296771.1
NM_001309861.1 132 Intron NP_001296790.1
NM_001309883.1 132 Intron NP_001296812.1
NM_016592.3 132 UTR 5 NP_057676.1
NM_080425.3 132 Intron NP_536350.2
NM_080426.3 132 Intron NP_536351.1
XM_017027812.1 132 Intron XP_016883301.1
XM_017027813.1 132 Intron XP_016883302.1
XM_017027814.1 132 Intron XP_016883303.1
XM_017027815.1 132 Intron XP_016883304.1
XM_017027816.1 132 Intron XP_016883305.1
XM_017027817.1 132 Intron XP_016883306.1
XM_017027818.1 132 Intron XP_016883307.1
XM_017027819.1 132 Intron XP_016883308.1
XM_017027820.1 132 Intron XP_016883309.1
XM_017027821.1 132 UTR 5 XP_016883310.1
XM_017027822.1 132 UTR 5 XP_016883311.1
Gene
GNAS-AS1
Gene Name
GNAS antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details