Product Details

SNP ID
rs56172300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58988433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATCTGTGGCTGGGAGCAGCCTC[A/G]GGGAGCTGGCCTTGGCACAAACAGG
Phenotype
MIM: 603169 MIM: 605297
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTSZ PubMed Links

Gene Details

Gene
CTSZ
Gene Name
cathepsin Z
There are no transcripts associated with this gene.

Gene
NELFCD
Gene Name
negative elongation factor complex member C/D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198976.2 Intron NP_945327.2

View Full Product Details