Product Details
- SNP ID
-
rs55893536
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:44942185 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTACTACAGAGTTCCAGGTGCAAAG[C/T]TGAAAAGGCATAAATGAAAGCACAG
- Phenotype
-
MIM: 616049
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PABPC1L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78973918] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PABPC1L
- Gene Name
- poly(A) binding protein cytoplasmic 1 like
- Gene
- TOMM34
- Gene Name
- translocase of outer mitochondrial membrane 34
View Full Product Details