Product Details

SNP ID
rs57034992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45607943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAAAATATGCAGAGCCCTCAG[G/T]TTGATGTAGCAGTTTATTTGACAAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
WFDC9 PubMed Links

Gene Details

Gene
WFDC9
Gene Name
WAP four-disulfide core domain 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147198.3 655 UTR 3 NP_671731.1

View Full Product Details