Product Details
- SNP ID
-
rs55915202
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:4221863 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGAGCGCGGTGAGGGCCAGCGGC[A/G]CTCCGGGGGGCGCGTCGCCCGAACT
- Phenotype
-
MIM: 104219
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ADRA1D
PubMed Links
Gene Details
- Gene
- ADRA1D
- Gene Name
- adrenoceptor alpha 1D
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000678.3 |
1434 |
Missense Mutation |
GCG,GTG |
A460V |
NP_000669.1 |
View Full Product Details