Product Details

SNP ID
rs55983705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:6041345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGCCCGTACAGAGGGTGCTGCC[G/T]GGCCGTGGCGTAGATGACCCCCACC
Phenotype
MIM: 608188
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CRLS1 PubMed Links

Gene Details

Gene
CRLS1
Gene Name
cardiolipin synthase 1
There are no transcripts associated with this gene.

Gene
LRRN4
Gene Name
leucine rich repeat neuronal 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152611.4 1136 Silent Mutation GGG,TGG G634W NP_689824.2
XM_011529183.2 1136 Missense Mutation XP_011527485.1

View Full Product Details