Product Details

SNP ID
rs56007823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:25388068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACTGACCTTCAGCCCTAACCAG[A/G]TCTTGTTTCCTATCATCTTGCTTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00158 PubMed Links

Gene Details

Gene
LINC00158
Gene Name
long intergenic non-protein coding RNA 158
There are no transcripts associated with this gene.

View Full Product Details