Product Details

SNP ID
rs55641247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:26837667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTCATGAGATAACACCCCTCCA[C/T]CATGGGACAGACACTTCAAGCTTCT
Phenotype
MIM: 605174
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS1 PubMed Links

Gene Details

Gene
ADAMTS1
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006988.4 3271 Missense Mutation GAT,GGT D939G NP_008919.3

View Full Product Details