Product Details

SNP ID
rs59988518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:14962939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATAATTTTTGATAGGTCATTTG[C/T]TAACCTGGTTCACAATTAAGAACAA
Phenotype
MIM: 602490
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NRIP1 PubMed Links

Gene Details

Gene
NRIP1
Gene Name
nuclear receptor interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003489.3 6071 UTR 3 NP_003480.2
XM_005261063.3 6071 UTR 3 XP_005261120.1
XM_005261065.3 6071 UTR 3 XP_005261122.1
XM_011529747.1 6071 UTR 3 XP_011528049.1
XM_011529748.2 6071 UTR 3 XP_011528050.1
XM_011529749.2 6071 UTR 3 XP_011528051.1
XM_011529751.2 6071 UTR 3 XP_011528053.1
XM_011529752.1 6071 UTR 3 XP_011528054.1
XM_017028473.1 6071 UTR 3 XP_016883962.1
XM_017028474.1 6071 UTR 3 XP_016883963.1
XM_017028475.1 6071 UTR 3 XP_016883964.1
XM_017028476.1 6071 UTR 3 XP_016883965.1

View Full Product Details