Product Details

SNP ID
rs56307674
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:34368805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTATTCTAGCTGGTGTGAGGTTA[C/T]CAATTTTTTTTAAACAAAAGTAATA
Phenotype
MIM: 603796
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KCNE2 PubMed Links
Additional Information
For this assay, SNP(s) [rs138091848] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KCNE2
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172201.1 Intron NP_751951.1
Gene
SMIM11A
Gene Name
small integral membrane protein 11A
There are no transcripts associated with this gene.

View Full Product Details