Product Details

SNP ID
rs56165916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:17820031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGACTTTTGTTTAAGCTATTCTG[G/T]TTTTTTTAGGGTATGGTTCTTGCAG
Phenotype
MIM: 607247
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C21orf91 PubMed Links

Gene Details

Gene
C21orf91
Gene Name
chromosome 21 open reading frame 91
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100420.1 Intron NP_001093890.1
NM_001100421.1 Intron NP_001093891.1
NM_017447.3 Intron NP_059143.3
Gene
CHODL
Gene Name
chondrolectin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204174.1 Intron NP_001191103.1
NM_001204175.1 Intron NP_001191104.1
NM_001204176.1 Intron NP_001191105.1
NM_001204177.1 Intron NP_001191106.1
NM_001204178.1 Intron NP_001191107.1
NM_024944.2 Intron NP_079220.2
XM_011529453.2 Intron XP_011527755.1
XM_011529457.2 Intron XP_011527759.1
XM_017028273.1 Intron XP_016883762.1

View Full Product Details