Product Details
- SNP ID
-
rs59152409
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:44304914 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGAGGCACTGTTCCTTTAGAGAC[A/G]GCGTTGCTTGCTCAAGTGGAGAAGC
- Phenotype
-
MIM: 607358
MIM: 171860
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AIRE
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201544793] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AIRE
- Gene Name
- autoimmune regulator
There are no transcripts associated with this gene.
- Gene
- PFKL
- Gene Name
- phosphofructokinase, liver type
View Full Product Details