Product Details

SNP ID
rs58520124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44941906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACGAGCTGCAGACAAGGCGAGTC[C/T]CTTCTGGCAGAGCTATGGAGTTCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM207A PubMed Links

Gene Details

Gene
FAM207A
Gene Name
family with sequence similarity 207 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316983.1 Intron NP_001303912.1
NM_001316984.1 Intron NP_001303913.1
NM_001316985.1 Intron NP_001303914.1
NM_001316986.1 Intron NP_001303915.1
NM_001316987.1 Intron NP_001303916.1
NM_001316988.1 Intron NP_001303917.1
NM_058190.3 Intron NP_478070.1
XM_017028481.1 Intron XP_016883970.1
XM_017028482.1 Intron XP_016883971.1
Gene
LINC01547
Gene Name
long intergenic non-protein coding RNA 1547
There are no transcripts associated with this gene.

View Full Product Details