Product Details

SNP ID
rs55637746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44990047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGAAAACAGCCGCTGTGATAAAC[C/G]CAAGGCCGAAAGCCGTCTCCGGGGA
Phenotype
MIM: 610259
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00163 PubMed Links

Gene Details

Gene
LINC00163
Gene Name
long intergenic non-protein coding RNA 163
There are no transcripts associated with this gene.

Gene
PICSAR
Gene Name
P38 inhibited cutaneous squamous cell carcinoma associated lincRNA
There are no transcripts associated with this gene.

View Full Product Details