Product Details

SNP ID
rs62222181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20081591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAATGTGCTTCTGGCTGGCTTG[C/T]CCAGTCTTAGACTCTCTTGTTGTAG
Phenotype
MIM: 609030
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGCR8 PubMed Links

Gene Details

Gene
DGCR8
Gene Name
DGCR8 microprocessor complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190326.1 Intron NP_001177255.1
NM_022720.6 Intron NP_073557.3
XM_006724268.2 Intron XP_006724331.1
Gene
MIR1306
Gene Name
microRNA 1306
There are no transcripts associated with this gene.

Gene
MIR3618
Gene Name
microRNA 3618
There are no transcripts associated with this gene.

View Full Product Details