Product Details

SNP ID
rs62235995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37644090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTGAATGGTGAATAACAGGTCA[C/T]TGTGCAAGGCCAGGCGCGGTGGCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101927051 PubMed Links

Gene Details

Gene
LOC101927051
Gene Name
uncharacterized LOC101927051
There are no transcripts associated with this gene.

Gene
SH3BP1
Gene Name
SH3 domain binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018957.3 Intron NP_061830.3

View Full Product Details