Product Details
- SNP ID
-
rs62228811
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:38863302 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATGATACTTGGGTGGGGAGGGGGC[A/G]CACTTAAGGGGAGATGACGGATGCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CBX6
PubMed Links
Gene Details
- Gene
- CBX6
- Gene Name
- chromobox 6
View Full Product Details