Product Details

SNP ID
rs58816224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29317075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCTAGATCGGGGGCTGGGGTCC[A/G]CTTTCACTCCTCTCCTCAGCCACTA
Phenotype
MIM: 602128 MIM: 602220
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAS2L1 PubMed Links

Gene Details

Gene
GAS2L1
Gene Name
growth arrest specific 2 like 1
There are no transcripts associated with this gene.

Gene
RASL10A
Gene Name
RAS like family 10 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006477.4 244 Intron NP_006468.1
XM_011529821.1 244 Intron XP_011528123.1
XM_011529822.1 244 UTR 5 XP_011528124.1
XM_011529823.1 244 Intron XP_011528125.1

View Full Product Details