Product Details

SNP ID
rs60825978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29481007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTCGGCCAGATCCAGGGCTCC[A/G]GCGCCGCGCAGGCGCAGATGCAGGC
Phenotype
MIM: 162230
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NEFH PubMed Links

Gene Details

Gene
NEFH
Gene Name
neurofilament heavy polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021076.3 Intron NP_066554.2
XM_011530200.2 Intron XP_011528502.1

View Full Product Details