Product Details

SNP ID
rs61098773
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29511618 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACGTGTTCCATGGCATGTTTGA[A/C]TTAGTAGCAGCAATGCTGATTAACA
Phenotype
MIM: 612733
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
THOC5 PubMed Links

Gene Details

Gene
THOC5
Gene Name
THO complex 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002877.1 Intron NP_001002877.1
NM_001002878.1 Intron NP_001002878.1
NM_001002879.1 Intron NP_001002879.1
NM_003678.4 Intron NP_003669.4
XM_005261797.1 Intron XP_005261854.1
XM_005261798.1 Intron XP_005261855.1
XM_005261799.1 Intron XP_005261856.1
XM_005261801.3 Intron XP_005261858.1
XM_006724347.1 Intron XP_006724410.1
XM_017029027.1 Intron XP_016884516.1

View Full Product Details