Product Details
- SNP ID
-
hCV89596649
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:75053441 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTGTAGTTTCTTTCTTTCCTCCT[C/T]TTTGGATATATTTTCTTTCTTTGCT
- Phenotype
-
MIM: 300135
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ABCB7
PubMed Links
Gene Details
- Gene
- ABCB7
- Gene Name
- ATP binding cassette subfamily B member 7
There are no transcripts associated with this gene.
View Full Product Details