Product Details
- SNP ID
-
rs56357230
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:66170731 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCATCACGAACCAGCCTCTGGACA[A/G]TGACATGTAGGTTTAATTTCTTGTG
- Phenotype
-
MIM: 300167
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HEPH
PubMed Links
Gene Details
- Gene
- HEPH
- Gene Name
- hephaestin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001130860.3 |
272 |
Missense Mutation |
AAT,AGT |
N57S |
NP_001124332.1 |
NM_001282141.1 |
272 |
Missense Mutation |
AAT,AGT |
N57S |
NP_001269070.1 |
NM_014799.3 |
272 |
Intron |
|
|
NP_055614.1 |
NM_138737.4 |
272 |
Missense Mutation |
AAT,AGT |
N108S |
NP_620074.3 |
XM_006724722.1 |
272 |
Missense Mutation |
AAT,AGT |
N57S |
XP_006724785.1 |
XM_011531073.2 |
272 |
Missense Mutation |
AAT,AGT |
N108S |
XP_011529375.1 |
XM_011531074.2 |
272 |
Missense Mutation |
AAT,AGT |
N108S |
XP_011529376.1 |
XM_011531075.2 |
272 |
Missense Mutation |
AAT,AGT |
N108S |
XP_011529377.1 |
XM_017029998.1 |
272 |
Missense Mutation |
AAT,AGT |
N108S |
XP_016885487.1 |
XM_017029999.1 |
272 |
Missense Mutation |
AAT,AGT |
N57S |
XP_016885488.1 |
XM_017030000.1 |
272 |
Missense Mutation |
AAT,AGT |
N57S |
XP_016885489.1 |
XM_017030001.1 |
272 |
Missense Mutation |
AAT,AGT |
N57S |
XP_016885490.1 |
XM_017030002.1 |
272 |
Missense Mutation |
AAT,AGT |
N57S |
XP_016885491.1 |
XM_017030003.1 |
272 |
Missense Mutation |
AAT,AGT |
N57S |
XP_016885492.1 |
View Full Product Details